Thermo Scientific™ single-use systems are commonly used to store and transport sterile process liquids, buffers, culture media, and bulk drug precursors. Our portfolio of shipping-validated outer support containers includes large volume stainless steel containers, plastic bins, and cylindrical drums.
These containers are designed to hold Thermo Scientific™ bioprocess containers (BPCs), and to support your transportation and liquid handling needs in biopharmaceutical and biomanufacturing applications.
For custom media or standard liquids needs, we routinely deliver liquid media and buffers in our BPCs in outer support containers to customers around the world.
Large Volume Stainless Steel Shipping Systems
Our standard stainless steel shippers for bioprocess containers and bags are designed to integrate with Envirotainer™ systems.
Key Features:
  • Simple, durable, ergonomic, and economic design constructed in 304 stainless steel
  • Suitable for both top and bottom discharge
  • Movable by forklift, pallet jack, or available carts
  • Adjustable dunnage system
  • Validated shipping configuration
  • Standard BPC chambers available
  • Compatible systems are available for temperature-controlled shipping in aircraft
Applications:
  • Sterile media and buffer storage and transport within and between facilities
  • Storage and transport of bioreactor harvest, sterile process liquids and buffers, and bulk drug precursors
AllpaQ Containers
AllpaQ™ containers are rigid plastic bins specifically created for the storage and shipment of liquids held in bioprocess containers.
Key Features:
  • Cleanable—Made of polypropylene with smooth, easy–to-clean surfaces
  • Reusable—Robust design and sturdy construction allows for multiple uses
  • Convenient—Collapsible walls fold down for easy storage when bins are not in use
  • Stackable—Easy storage both empty and full
  • Bottom drain—True bottom drain options are available
Applications:
  • Shipping and transportation
  • In-house storage
Standard Cylindrical Drums
Cylindrical drums are economic, validated shipping systems for BPCs.
Key Features:
  • Constructed of linear low density polyethylene (LLDPE)
  • Available in a range of volumes
  • Available in top- or bottom-drain options
  • Support a range of standard BPC bags and chamber designs
Applications:
  • Shipping and transportation
  • In-house applications such as harvesting and storage in non-critical areas
BioProduction resources
Have questions about single-use technologies, Gibco™ media, POROS™ chromatography resins, Applied Biosystems™ SEQ rapid contaminant tests, or our Cell Therapy Systems products?  Click below to access product brochures, training videos, mobile apps and more.