The discovery of cancer stem cells in certain tumor types launched a new discipline within cancer research. Many scientists are now looking to identify and characterize populations of tumor cells which have acquired stem cell–like properties as potential sources of metastasis and therapeutic resistance.
Thermo Fisher Scientific offers solutions for cancer stem cell research—from identification and culturing to genomic and functional analysis.
What are cancer stem cells?
Cancer stem cells are a small subpopulation of tumor cells with stem-like properties that drive tumor initiation, maintenance, metastasis, and therapy resistance.
Key characteristics of cancer stem cells
  • Self-renewal—ability to continuously generate more CSCs
  • Tumor-initiating capacity—can form new tumors when transplanted in vivo
  • Differentiation potential—give rise to diverse cell types within a tumor
  • Therapy resistance—more resilient to chemotherapy and radiation
  • Metastatic competence—contribute to tumor spread and recurrence
  • Distinct molecular markers—expression of surface and intracellular markers (varies by cancer type)
Table 1. Common cancer stem cell markers (examples).
Cancer typeFrequently studied markers
BrainCD133, Nestin, SOX2
BreastCD44high/CD24low, ALDH1
ColonCD133, CD44, EpCAM
PancreaticCD44, CD24, ESA
LeukemiaCD34+/CD38-
Table 2. Methods to identify cancer stem cells.
CategoryExamplesWhat it shows
Cell-surface marker detectionFlow cytometry using CD133, CD44/CD24, etc.Enrichment of stem-like subpopulations
Functional assaysSphere formation assays (e.g., tumorspheres, neurospheres)Self-renewal and survival in non-adherent culture
Stem-ness reporter assaysSOX2/OCT4 promoter reporters, ALDH activity assaysStem-cell gene expression/activity
In vivo tumor-initiating assaysXenografts in immunodeficient miceGold-standard proof of tumor-initiating ability
Drug-resistance assaysResistance to chemo/radiation, efflux pump assaysTherapy-resistant phenotype
Cancer stem cell workflow
decorative
decorative
Cancer stem cell culture
For your cancer stem cell needs, use trusted and widely referenced cell culture media, reagents, cells, and sera.
Recommended products & resources
Gibco cell culture products
Primary cell culture systems
Stem cell research
Recombinant proteins for stem cell culture
AlgiMatrix 3D Cell Culture System
sequencing and qPCr systems and equipment for cancer stem cell characterization
Cancer stem cell characterization
Characterize the genomic events of cancer stem cells driving regenerative properties with highly sensitive sequencing and qPCR technologies. Thermo Fisher Scientific portfolio supports a wide range of throughput and application needs for genomic analysis.
  • Applied Biosystems Genetic Analyzers—exceptional for sequencing and fragment analysis.
  • Applied Biosystems and Invitrogen products—excellent PCR systems and portfolios
Recommended products & resources
Next Generation Sequencing (NGS)
Oligonucleotides, Primers, Probes, & Genes
Sanger Sequencing and Fragment Analysis by Capillary Electrophoresis
Attune NxT Flow Cytometer for cancer stem cell functional assays
Cancer stem cell functional assays
Detect cancer stem cells quickly without loss of sensitivity with the Invitrogen Attune NxT Flow Cytometer. Uncover diverse cell processes in live cells with Invitrogen reagents and assays.
Recommended products & resources
Attune Flow Cytometers
Flow Cytometry Solutions
Cell Function Assays
decorative
decorative
Cancer stem cell culture
For your cancer stem cell needs, use trusted and widely referenced cell culture media, reagents, cells, and sera.
Recommended products & resources
Gibco cell culture products
Primary cell culture systems
Stem cell research
Recombinant proteins for stem cell culture
AlgiMatrix 3D Cell Culture System
sequencing and qPCr systems and equipment for cancer stem cell characterization
Cancer stem cell characterization
Characterize the genomic events of cancer stem cells driving regenerative properties with highly sensitive sequencing and qPCR technologies. Thermo Fisher Scientific portfolio supports a wide range of throughput and application needs for genomic analysis.
  • Applied Biosystems Genetic Analyzers—exceptional for sequencing and fragment analysis.
  • Applied Biosystems and Invitrogen products—excellent PCR systems and portfolios
Recommended products & resources
Next Generation Sequencing (NGS)
Oligonucleotides, Primers, Probes, & Genes
Sanger Sequencing and Fragment Analysis by Capillary Electrophoresis
Attune NxT Flow Cytometer for cancer stem cell functional assays
Cancer stem cell functional assays
Detect cancer stem cells quickly without loss of sensitivity with the Invitrogen Attune NxT Flow Cytometer. Uncover diverse cell processes in live cells with Invitrogen reagents and assays.
Recommended products & resources
Attune Flow Cytometers
Flow Cytometry Solutions
Cell Function Assays
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.