Transcription factors are proteins that bind to specific DNA sequences and regulate transcription  from DNA to mRNA by either activating or inhibiting RNA polymerase. These transcription factors contain more than one DNA-binding domain which allows the protein to bind to specific DNA sequences near the genes that they regulate.
Invitrogen offers a wide selection of highly specific primary antibodies for studying transcription factors that have been validated for multiple applications.
Primary Antibody Search Tool