a gloved scientist's hand holding a microcentrifuge tube containing pink-colored liquid that is being withdrawn using a pipette  
This protocol provides basic instructions to perform a fluorescence-based assay to detect functional mictochondria. Healthy mitochondrial membranes maintain a difference in electrical potential between the interior and exterior of the organelle, referred to as a membrane potential. Tetramethylrhodamine, methyl ester (TMRM) is a cell-permeant dye that accumulates in active mitochondria with intact membrane potentials. If the cells are healthy and have functioning mitochondria, the signal will be bright. Upon loss of the mitochondrial membrane potential, TMRM accumulation will cease and the signal will dim or disappear.
Download this protocol
Reminder
This assay is for live cells.
What you need
  • Live cells
  • Complete medium (whatever you are growing your cells in)
  • Tetramethylrhodamine, methyl ester (TMRM)
  • Phosphate-buffered saline (PBS), or any other saline-based buffer
  • TRITC filter set for your fluorescence microscope
Functional mitochondrial staining—basic labeling protocol
This can be a protocol tip and should read: TMRM is often supplied as a powder. It’s useful to make a stock solution in DMSO and store it at –20°C (for example, add 5 mL DMSO to 25 mg of TMRM to make a 10 mM stock solution). Once you’re ready to use your stock, because TMRM is used at a low working concentration, you will need to make intermediate dilutions.
  • To make 50 µM intermediate dilution of TMRM → 1 µL 10 mM TMRM + 200 µL complete medium
  • To make 250 nM staining solution of TMRM → 5 µL 50 µM TMRM + 1 mL complete medium
  • The volumes given in this protocol are good for a single well in a 6-well vessel or a single 35 mm vessel
1 Prepare 1 mL of 250 nM TMRM staining solution in complete medium.If you are optimizing for dye concentration, you will need to prepare a staining solution for each concentration you want to test.
2 Remove media from live cells.
3 Add the TMRM staining solution.
4 Incubate for 30 minutes at 37°C.
5 Wash 3 times with PBS (or other clear buffer). Learn how to wash 
6 Image using TRITC filter.