Cell Isolation–Use Your Own Antibody
Dynabeads products are used for the isolation of a range of different species-specific cell populations. Below you can read a selection of publications detailing the application of Dynabeads products in addition to links to related products.
For further information, please contact our Technical Service.
Dynabead products in the literature—select publications:
See also:
- Dynabeads FlowComp Flexi—combine with any antibody and obtain bead-free cells
- Cell Isolation using Biotinylated Antibodies—isolate any cell type from any sample from any species
- List of Dynabeads for use with your own antibody—a list of products by starting sample and related benefits
Dynabeads products in the literature
Read about practical applications of Dynabeads products to isolate cell types in a species-specific manner. These publications highlight the utility of Dynabeads products in positive cell isolation, negative cell isolation and depletion of cell types from mixed cell populations.
Product | Starting sample | Benefits |
---|---|---|
Dynabeads FlowComp Flexi | Single Cell Suspensions | Combine with your own antibody of choice to isolate any cell type from any species. This kit provides everything you need for labeling of antibodies and subsequent cell isolation. Beads are removed after isolation—you obtain pure and viable cells suitable for any downstream application. |
CELLection Biotin Binder | Whole blood, MNC | Use with any biotinylated antibody to positively isolate cells from any species. The beads are removed after isolation—you obtain pure and viable cells for use in any application. [More info] |
Dynabeads Biotin Binder | Whole blood, MNC | Isolate any cell from any species. This kit bind all biotinylated antibodies and is our most flexible secondary product for cell isolation or depletion. [More info] |
Product | Starting sample | Benefits |
---|---|---|
Dynabeads Sheep anti-Rat IgG | Whole blood, MNC | Isolate any cell type from any species except rat. For use with your primary IgG rat antibody of choice. Ideal for positive isolation, negative isolation or depletion of cells. |
Dynabeads Pan Mouse IgG | Whole blood, MNC | A flexible secondary product for cell isolation or depletion. It will bind all subclasses of mouse IgG and is Fc specific. Requires less antibody and has shown no cross reactivity to human, rat, hamster, goat, rabbit and guinea pig IgG. |
CELLection Pan Mouse IgG | Whole blood, MNC | Use with any mouse IgG to positively isolate cells from any species EXCEPT mouse. Beads can be removed after isolation to leave pure and viable cells for use in any application. |
Dynabeads Sheep anti-Mouse IgG | Whole blood, MNC | Isolate any cell type from any species except mouse. For use with your primary mouse IgG1, IgG2a or IgG2b antibody of choice. Ideal for positive isolation, negative isolation or depletion of cells. Also available in clinical research grade as Dynabeads ClinExVivo Sheep anti-Mouse IgG. |
Dynabeads Goat anti-Mouse IgG | Whole blood, MNC | Isolate any cell type from any species except mouse. For use with your primary IgG mouse antibody of choice. Ideal for positive isolation, negative isolation or depletion of cells. |
Dynabeads Rat anti-Mouse IgG1 | Whole blood, MNC | Isolate any cell type from any species except mouse. For use with your primary mouse IgG1 antibody of choice. Ideal for positive isolation, negative isolation or depletion of cells. |
Dynabeads Rat anti-Mouse IgM | Whole blood, MNC | Isolate any cell type from any species except mouse. For use with your primary mouse IgM antibody of choice. Ideal for positive isolation, negative isolation or depletion of cells. |
Dynabeads Sheep anti-Rabbit IgG | Whole blood, MNC | Isolate any cell type from any species except rabbit. For use with your primary rabbit antibody of choice. This product can be used for eucaryotic cells. |
Product | Starting sample | Benefits |
---|---|---|
Dynabeads M-450 Epoxy |
Whole blood, MNC | Hydrophobic naked beads with glycidyl ether (epoxy) reactive groups. Ideal for coupling of antibodies for cell applications. Coupling reactions can be performed at neutral pH, with 24 hour incubation at room temperature. Also available in clinical research grade as Dynabeads ClinExVivo Epoxy. |
Dynabeads M-450 Tosylactivated |
Whole blood, MNC | Hydrophobic naked beads with p-toluenesulphonyl (tosyl) reactive groups. For coupling of antibodies for cell applications. Coupling reactions can be performed at neutral pH although high pH and incubation at 37°C will promote covalent binding. |
Product | Starting sample | Benefits |
---|---|---|
Dynabeads FlowComp Flexi | Single Cell Suspensions | Combine with your own antibody of choice to isolate any cell type from any species. This kit provides everything you need for labeling of antibodies and subsequent cell isolation. Beads are removed after isolation—you obtain pure and viable cells suitable for any downstream application. |
CELLection Biotin Binder | Whole blood, MNC | Use with any biotinylated antibody to positively isolate cells from any species. The beads are removed after isolation—you obtain pure and viable cells for use in any application. [More info] |
Dynabeads Biotin Binder | Whole blood, MNC | Isolate any cell from any species. This kit bind all biotinylated antibodies and is our most flexible secondary product for cell isolation or depletion. [More info] |
Product | Starting sample | Benefits |
---|---|---|
Dynabeads Sheep anti-Rat IgG | Whole blood, MNC | Isolate any cell type from any species except rat. For use with your primary IgG rat antibody of choice. Ideal for positive isolation, negative isolation or depletion of cells. |
Dynabeads Pan Mouse IgG | Whole blood, MNC | A flexible secondary product for cell isolation or depletion. It will bind all subclasses of mouse IgG and is Fc specific. Requires less antibody and has shown no cross reactivity to human, rat, hamster, goat, rabbit and guinea pig IgG. |
CELLection Pan Mouse IgG | Whole blood, MNC | Use with any mouse IgG to positively isolate cells from any species EXCEPT mouse. Beads can be removed after isolation to leave pure and viable cells for use in any application. |
Dynabeads Sheep anti-Mouse IgG | Whole blood, MNC | Isolate any cell type from any species except mouse. For use with your primary mouse IgG1, IgG2a or IgG2b antibody of choice. Ideal for positive isolation, negative isolation or depletion of cells. Also available in clinical research grade as Dynabeads ClinExVivo Sheep anti-Mouse IgG. |
Dynabeads Goat anti-Mouse IgG | Whole blood, MNC | Isolate any cell type from any species except mouse. For use with your primary IgG mouse antibody of choice. Ideal for positive isolation, negative isolation or depletion of cells. |
Dynabeads Rat anti-Mouse IgG1 | Whole blood, MNC | Isolate any cell type from any species except mouse. For use with your primary mouse IgG1 antibody of choice. Ideal for positive isolation, negative isolation or depletion of cells. |
Dynabeads Rat anti-Mouse IgM | Whole blood, MNC | Isolate any cell type from any species except mouse. For use with your primary mouse IgM antibody of choice. Ideal for positive isolation, negative isolation or depletion of cells. |
Dynabeads Sheep anti-Rabbit IgG | Whole blood, MNC | Isolate any cell type from any species except rabbit. For use with your primary rabbit antibody of choice. This product can be used for eucaryotic cells. |
Product | Starting sample | Benefits |
---|---|---|
Dynabeads M-450 Epoxy |
Whole blood, MNC | Hydrophobic naked beads with glycidyl ether (epoxy) reactive groups. Ideal for coupling of antibodies for cell applications. Coupling reactions can be performed at neutral pH, with 24 hour incubation at room temperature. Also available in clinical research grade as Dynabeads ClinExVivo Epoxy. |
Dynabeads M-450 Tosylactivated |
Whole blood, MNC | Hydrophobic naked beads with p-toluenesulphonyl (tosyl) reactive groups. For coupling of antibodies for cell applications. Coupling reactions can be performed at neutral pH although high pH and incubation at 37°C will promote covalent binding. |
Related pages
Resources
Literature
For Research Use Only. Not for use in diagnostic procedures.