Invitrogen EVOS M7000 Imaging System
Powerful, flexible, fast, automated Imaging system
The EVOS M7000 Imaging System replaces the EVOS FL Auto 2. It features high-resolution CMOS cameras (mono and color) with improved resolution and sensitivity. It performs rapid autofocusing, image acquisition and processing of large data. Compatible with the accessories of FL Auto 2, including the EVOS Onstage Incubator for easy and simplified live-cell analysis, image tiling, and z-stacking
Request a demo  Request a quote
Watch and make way for More with Invitrogen EVOS M7000 Imaging System
Learn why EVOS M7000 Imaging System makes way for More
  • Speed—scan a 96-well plate in 3 fluorescence channels in less than 5 minutes
  • Flexibility—customize the system with more than 20 user-changeable LED light cubes, dual cameras (monochrome and color), a variety of objectives ranging from 1.25x to 100x, and multiple vessel holders
  • Time-lapse live-cell imaging—onstage incubator option for precise control of temperature, humidity, and gases for normoxic or hypoxic conditions allows a wide range of biological studies under physiological conditions
  • Two cameras, no compromises—all systems come with two cameras: a dedicated high-sensitivity monochrome camera optimized for fluorescence imaging and quantitation, and a dedicated high-resolution color camera optimized for colorimetric imaging
  • Area view—move rapidly and seamlessly between single-field mode and low- and high-magnification scan modes to easily define and capture the area of interest
  • Automation—time-saving features such as autofocus, rapid stage movement, and automated routines help reduce time to complete experiments, allowing high throughput, high data quality, and improved experimental reproducibility
  • Data analysis—extensive quantitative imaging and statistical analysis in combination with Invitrogen Celleste Image Analysis Software, an optional advanced software package offering powerful tools for image segmentation and classification that can be used for cell counting and measuring changes
The EVOS M7000 Imaging System offers you these important advantages:
Images captured using the EVOS M7000 Imaging System
Figure 1. HeLa cells expressing Mitochondria RFP were plated on a 6-well Falcon plate. Cells were then stained with NucBlue Live and imaged in Live Cell Imaging Solution using 40x objective (Cat. No. AMEP4983).
Figure 2. EdU labeling combined with Movat's Pentachrome stain shows compatibility of DAB with multiple color pentachrome staining. Rat intestinal crypts shows proliferating cell locally abundant near the base of the crypts and diminishing with distance. Proliferating cells (brown) signal are visible against Movat's Pentachrome staining consisting of Verhoeff-Van Gieson’s staining of elastin (black) and nuclei (purplish gray), alcoholic saffron staining of collagen (yellow), Alcian Blue staining of mucins marking the goblet cells (blue), and Crocein Scarlet–Acid Fuchsin staining of muscle and red blood cells (red).
System and hardware specifications
Optics: Infinity‐corrected optical system; RMS‐threaded objectives with 45 mm parfocal distance
Imaging mode: Fluorescence, brightfield, color brightfield, and phase contrast
Illumination: Five position chamber for 4 fluorescence light cubes plus brightfield. Light cubes with integrated hard coated filter set and LED light source with >50,000 hour life. Broad selection of standard and specialty light cubes.
Imaging methods: Single color, multi-color, area scan with montage or tile-stitch, time lapse, Z-stacking, movie capture
Objective capacity: 5‐position turret
Objectives (not included): Wide selection of high‐quality LWD and coverslip‐corrected objectives
Condenser: 60 mm long working distance condenser, 4 position turret with a clear aperture and 3 phase annuli.
Stage: Motorized X/Y scanning stage. Travel range 120 mm x 80 mm with sub‐micron resolution, drop‐in inserts to receive vessel holders and lock down holders to fix sample in place during long scans.
Focus Mechanism: Automated focus mechanism with sub‐micron resolution
LCD display: 23" high‐resolution touch screen color monitor (also fully controllable via mouse); 1920x1080 pixel resolution.
Cameras:
  • High-sensitivity 3.2 MP (2048 x 1536) monochrome CMOS sensor with 3.45 µm pixel resolution
  • High-sensitivity 3.2 MP (2048 x 1536) color CMOS sensor with 3.45 µm pixel resolution
Computer: External Dell PC with an Intel Core i7-8700 processor, 32 GB DDR4 RAM, 512 GB PCIe SSD and NVIDIA Quadro P1000 with 4GB discrete video graphics running Windows® 10, designed to operate with touchscreen monitor and microscope
Captured Images: 16‐bit RAW monochrome: TIFF, PNG
8-bit TIFF, PNG, JPG
Movies and time‐lapse: AVI, WMV
Output ports: Computer: 1 x USB 3.1 Gen 2 Type-C; 5 x USB 3.1 Gen 1 Type-A; 4 x USB 2.0 Type-A; 1 Serial; 2 DisplayPorts 1.2; 1 RJ-45; 2 PS/2; 1 UAJ; 1 Line-out
Networking capability: Connection through Windows/SMB network via an Ethernet cable connection
Power supply: 24 V AC adapter with country‐specific power cords
Dimensions (LxWxH) 18x14x13 inches
457x330x356 mm
Weight: 16/35 (kg/lb)
Images captured using the EVOS M7000 Imaging System
Figure 1. HeLa cells expressing Mitochondria RFP were plated on a 6-well Falcon plate. Cells were then stained with NucBlue Live and imaged in Live Cell Imaging Solution using 40x objective (Cat. No. AMEP4983).
Figure 2. EdU labeling combined with Movat's Pentachrome stain shows compatibility of DAB with multiple color pentachrome staining. Rat intestinal crypts shows proliferating cell locally abundant near the base of the crypts and diminishing with distance. Proliferating cells (brown) signal are visible against Movat's Pentachrome staining consisting of Verhoeff-Van Gieson’s staining of elastin (black) and nuclei (purplish gray), alcoholic saffron staining of collagen (yellow), Alcian Blue staining of mucins marking the goblet cells (blue), and Crocein Scarlet–Acid Fuchsin staining of muscle and red blood cells (red).
System and hardware specifications
Optics: Infinity‐corrected optical system; RMS‐threaded objectives with 45 mm parfocal distance
Imaging mode: Fluorescence, brightfield, color brightfield, and phase contrast
Illumination: Five position chamber for 4 fluorescence light cubes plus brightfield. Light cubes with integrated hard coated filter set and LED light source with >50,000 hour life. Broad selection of standard and specialty light cubes.
Imaging methods: Single color, multi-color, area scan with montage or tile-stitch, time lapse, Z-stacking, movie capture
Objective capacity: 5‐position turret
Objectives (not included): Wide selection of high‐quality LWD and coverslip‐corrected objectives
Condenser: 60 mm long working distance condenser, 4 position turret with a clear aperture and 3 phase annuli.
Stage: Motorized X/Y scanning stage. Travel range 120 mm x 80 mm with sub‐micron resolution, drop‐in inserts to receive vessel holders and lock down holders to fix sample in place during long scans.
Focus Mechanism: Automated focus mechanism with sub‐micron resolution
LCD display: 23" high‐resolution touch screen color monitor (also fully controllable via mouse); 1920x1080 pixel resolution.
Cameras:
  • High-sensitivity 3.2 MP (2048 x 1536) monochrome CMOS sensor with 3.45 µm pixel resolution
  • High-sensitivity 3.2 MP (2048 x 1536) color CMOS sensor with 3.45 µm pixel resolution
Computer: External Dell PC with an Intel Core i7-8700 processor, 32 GB DDR4 RAM, 512 GB PCIe SSD and NVIDIA Quadro P1000 with 4GB discrete video graphics running Windows® 10, designed to operate with touchscreen monitor and microscope
Captured Images: 16‐bit RAW monochrome: TIFF, PNG
8-bit TIFF, PNG, JPG
Movies and time‐lapse: AVI, WMV
Output ports: Computer: 1 x USB 3.1 Gen 2 Type-C; 5 x USB 3.1 Gen 1 Type-A; 4 x USB 2.0 Type-A; 1 Serial; 2 DisplayPorts 1.2; 1 RJ-45; 2 PS/2; 1 UAJ; 1 Line-out
Networking capability: Connection through Windows/SMB network via an Ethernet cable connection
Power supply: 24 V AC adapter with country‐specific power cords
Dimensions (LxWxH) 18x14x13 inches
457x330x356 mm
Weight: 16/35 (kg/lb)