HCS DNA Damage Assays
In mammalian cells, a double-strand break (DSB) in genomic DNA is potentially lethal to a cell, and one of the responses to DSB formation as part of the repair process is phosphorylation of histone H2AX.
See product information for DNA damage assays
The HCS DNA Damage Kit
DNA damage can be measured as an indicator of genotoxicity using an antibody against phosphorylated H2AX. By combining specific antibody-based detection of DNA damage with a cytotoxicity indicator, both parameters can be measured simultaneously in the same cell.
The HCS DNA Damage Kit uses a secondary antibody conjugate to detect phosphorylated H2AX, Image-iT™ DEAD Green™ dye to detect cytotoxicity, and Hoechst™ 33342 to label nuclei in both live and dead cells. The kit is compatible with additional multiplexing steps, and results can be analyzed using automated image analysis techniques.
  HCS DNA Damage Kit
Using the HCS DNA Damage Kit, researchers can detect and quantitate DNA damage and changes in cell permeability.
Properties of the HCS DNA Damage Kit
  HCS DNA Damage Kit
Readout Combined genotoxicity and cytotoxicity detection compatible with automated image analysis in HCS
Common filter set DAPI FITC TRITC
Reporter Hoechst 33342 stain Image-iT DEAD Green dye Alexa Fluor 555 dye
Ex/Em (nm) 350/461 488/515 555/565
Live-cell compatible No Yes No
Added to growth media No Yes No
Formaldehyde fixable Cells are labeled after fixation Yes Cells are labeled after fixation
Multiplexing Can be multiplexed with additional detection reagents
Signal-to-noise 4 stars 5 stars 5 stars
Photostability 4 stars 5 stars 5 stars
Platform Imaging, HCS
Bibliography Citations
Format 1 kit (2 x 96-well plates)
Cat. No. H10292