The ArrayScan™ Live Cell Module provides complete quantitative, kinetic, live-cell imaging and analysis for Thermo Scientific™ ArrayScan™ HCA Readers. Study the dynamics of intracellular molecular interactions and cell motion in an environment similar to that of a tissue culture incubator. The ArrayScan™ Live Cell Module offers full control of temperature, carbon dioxide, and humidity levels to maintain cell health and viability, helping to ensure more reliable results.
Request a quote
Features
  • Full environmental control—complete environmental control (CO2, humidity, and temperature) with full reporting of CO2 and temperature over short or extended periods of time
  • Kinetic capabilities—cell tracking with powerful kinetic analysis software provides numerous motility and kinetic calculations at the cell and well levels
  • Focus on cell health—Zeiss™ Definite Focus is a hardware-based autofocus system that maintains focus and experimental reliability while supporting long-term imaging of live cells; phototoxicity is reduced by substituting software focusing with less harmful near-IR LED technology
Specifications
Autofocus system Hardware-based Zeiss™ Definite Focus autofocus system
Compatibility
Thermo Scientific™ ArrayScan™ HCA Readers
CO2 mixed gas Software controlled; 0 to 10% saturation with less than 0.1% variation measured across a plate; CO2 saturation recorded and reported through software at each time point
External control Module: D x W x H: 56 x 33 x 36 cm control unit for temperature and CO2 control
Relative humidity Maintains greater than 90% relative humidity
Software Thermo Scientific™ HCS Studio™ Cell Analysis Software
Temperature control Software controlled—ambient to 45°C; temperature variation is less than 0.5°C over 48 hours; temperature recorded and reported through the software at each time point
Ordering Information
Catalog # Product Quantity  
N010114A Thermo Scientific™ ArrayScan™ Live Cell Module 1 each Request a quote