Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
Request a quote
Objective lenses for ArrayScan™ and CellInsight™ platforms
Magnification Objective type Compatible instrument Catalog #
Zeiss 63x (0.75NA) LD Plan-Neofluar ArrayScan™ HCA Readers N010251
Zeiss 2.5x (0.075NA) Plan-Neofluar ArrayScan™ HCA Readers N010250
Zeiss 40x (0.75NA) Plan-Neofluar ArrayScan™ HCA Readers N010099
Zeiss 20x (0.8NA) Plan-Apochromat ArrayScan™ HCA Readers N010098
Zeiss 40x (0.5NA) LD A-Plan ArrayScan™ HCA Readers N010097
Olympus 1.25x (0.04NA) PlanApo N ArrayScan™ HCA Readers
(not to be used with confocal)
N010125
Olympus 2x (0.08NA) PlanApo CellInsight™ HCS Platforms NX1002X
Olympus 4x (0.16NA) U Plan S-Apo CellInsight™ HCS Platforms NX1004X