photo of the Slideport Slideholder, showing 4 slides inserted  
Image slides on your Thermo Scientific™ High Content Instrument using Thermo Scientific™ Slideport™ Slide Inserts.
Request a quote
Slideport™ Slide Inserts have been tested in imaging a wide range of sample types including fixed and live cells, tissue sections, tissue microarrays, and beads. Slideport™ Slide Inserts allow you to image slides as if you were imaging a standard microplate—any microscope or plate reader that accepts standard microplates will also accept Slideport™ Slide Inserts. You can easily stack slides and automate imaging, and Slideport™ Slide Inserts can even be used at a variety of temperatures.
Features
  • Two skirt heights are available to accommodate a variety of readers and objectives
  • Any microscope or plate reader that accepts standard microplates will also accept Slideport™ Slide Inserts
  • Image a wide range of sample types including fixed and live cells, tissue sections, tissue microarrays, and beads
  • Automate imaging of your slides by utilizing the Slideport™ Slide Inserts automation compatibility
  • Easily stack slides for automated imaging with or without lids
  • Slideport™ Slide Inserts can be utilized at a variety of temperatures from 4–37°C, allowing for flexibility of storage and imaging conditions
Ordering Information
Product Quantity Catalog #  
Thermo Scientific™ Slideport™ Slide Inserts
1 insert N0101111 Request a quote
Thermo Scientific™ Slideport™ Slide Inserts
3 inserts N0101113 Request a quote
Thermo Scientific™ Slideport™ Slide Inserts
5 inserts N0101115 Request a quote