With the Thermo Scientific CellInsight CX7 LZR high-content instrument, you can take advantage of the entire fluorescent spectrum to optimize your assay—and multiplex your components to ask more in-depth biological questions. Use the tables below to select reagents for each channel; they are divided into application areas, including products for segmentation, cell structure, and more.
  • Select the Fixed cells or Live/fixable cells tab to see a reagent selection that matches your detection protocol.
  • Click on individual product links (within colored cells) to see product details including spectral profiles, images, sample data sets, citations, and cell-based protocols.
  • Click on an application area to see selection guides for additional fluorescence-based tools and assays for imaging.
Fixed cell labeling
Blue Green/Yellow Orange Red Deep Red Near IR
Alexa Fluor
405
Alexa Fluor
488
Alexa Fluor
532
Alexa Fluor
555
Alexa Fluor
594
Alexa Fluor
647
Alexa Fluor
750
Alexa Fluor
790
  Goat anti-chicken IgY   Goat anti-chicken IgY Goat anti-chicken IgY Goat anti-chicken IgY    
Goat anti-mouse IgG Goat anti-mouse IgG Goat anti-mouse IgG Goat anti-mouse IgG Goat anti-mouse IgG Goat anti-mouse IgG Goat anti-mouse IgG Goat anti-mouse IgG
Goat anti-rabbit IgG Goat anti-rabbit IgG Goat anti-rabbit IgG Goat anti-rabbit IgG Goat anti-rabbit IgG Goat anti-rabbit IgG Goat anti-rabbit IgG Goat anti-rabbit IgG
  Goat anti-rat IgG   Goat anti-rat IgG Goat anti-rat IgG Goat anti-rat IgG    
  Goat anti-guinea pig IgG   Goat anti-guinea pig IgG Goat anti-guinea pig IgG Goat anti-guinea pig IgG    
  Goat anti-human IgG   Goat anti-human IgG Goat anti-human IgG Goat anti-human IgG    
  Donkey anti-goat IgG   Donkey anti-goat IgG Donkey anti-goat IgG Donkey anti-goat IgG   Donkey anti-goat IgG
  Donkey anti-mouse IgG   Donkey anti-mouse IgG Donkey anti-mouse IgG Donkey anti-mouse IgG   Donkey anti-mouse IgG
              Donkey anti-rabbit IgG
  Donkey anti-rat IgG     Donkey anti-rat IgG      
  Rabbit anti-goat IgG   Rabbit anti-goat IgG Rabbit anti-goat IgG Rabbit anti-goat IgG   Rabbit anti-goat IgG
  Rabbit anti-mouse IgG   Rabbit anti-mouse IgG Rabbit anti-mouse IgG Rabbit anti-mouse IgG   Rabbit anti-mouse IgG
  Rabbit anti-rat IgG     Rabbit anti-rat IgG      
  Chicken anti-goat IgG     Chicken anti-goat IgG Chicken anti-goat IgG    
  Chicken anti-mouse IgG     Chicken anti-mouse IgG Chicken anti-mouse IgG    
  Chicken anti-rabbit IgG     Chicken anti-rabbit IgG Chicken anti-rabbit IgG    
  Chicken anti-rat IgG     Chicken anti-rat IgG Chicken anti-rat IgG    
Live/fixable cell labeling
Violet Blue Green/Yellow Orange Red Deep Red
BFP CFP GFP
emGFP
YFP RFP
mOrange
DsRed
mCherry