Comparable to Brilliant Violet 605 conjugates
Comparable to Brilliant Violet 605 conjugates
Invitrogen eBioscience Super Bright 600 antibody conjugates for flow cytometry provide:
Multiplexing with Super Bright 600 antibody conjugates
Figure 1. Staining performance and post-fixation stability. (A) Comparison of mouse splenocytes stained with CD8a antibody conjugated to either Super Bright 600 (red, Cat. No. 63-0081-82) or Brilliant Violet™ 605 conjugate (gray), at the same antibody concentration. (B) Mouse splenocytes were stained with CD45R antibody conjugated to Super Bright 600 (Cat. No. 63-0452-82) and either left unfixed (red), or were fixed in Invitrogen eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) for 30 minutes (blue), 24 hours (orange), or 3 days (green).
Multiplexing compatibility Buffer Multiplexing considerations
Multiplexing with 1 Super Bright antibody conjugate Standard buffers applicable No special buffer required when only one Super Bright antibody conjugate is used in a panel
Multiplexing with 2 or more Super Bright antibody conjugates Super Bright Staining Buffer Special staining buffer is required prior to addition of Super Bright conjugates to reduce non-specific dye-dye interactions
Multiplexing with 1 or more Brilliant Violet™ antibody conjugates Super Bright Staining Buffer Special staining buffer is required to reduce non-specific dye-dye interactions. Use of the Super Bright Staining Buffer is recommended, but the similar Brilliant Stain Buffer can be substituted
Viability stain options Product Multiplexing considerations
Fixable LIVE/DEAD fixable dead cell stain kits No compatibility concerns
Non-fixable SYTOX non-fixable dead cell stains
Ready Flow Ready-to-use viability reagents
No compatibility concerns
Compatible with all Ready Flow reagents for viability
Product Multiplexing considerations
UltraComp eBeads microspheres UltraComp eBeads microspheres are compatible but OneComp eBeads are not compatible with violet lasers; the AbC Total Antibody Compensation Bead Kit is also compatible with the Super Bright antibody conjugates.
Staining Target Product Multiplexing considerations
Cytosolic staining
(cytokines)
Intracellular Fixation & Permeabilization Buffer Set No compatibility concerns
Nuclear staining
(transcription factors) 
Foxp3/Transcription Factor Staining Buffer Set No compatibility concerns
Support
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.