Search
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| 3 | 633–640 | 785LP | 637 | 800 | (in buffer) 2 | flow cytometry |
Invitrogen NovaFluor Red 800 dye is a Red-fluorescent dye with a unique emission spectrum when excited by the 633–640 nm laser. It fits in space after NovaFluor Red 755, a spectral space that is currently not accessible by other Invitrogen spectral dyes potentially allowing the detection of an additional marker in a complex multicolor panel.
NovaFluor Red 800 dye should generally be paired with low to moderately expressed antigens in multicolor panels. The macromolecule-based NovaFluor Red 800 dye is less photostable than typical NovaFluor dyes, but still remains at least as stable as protein tandem fluorophores. Use NovaFluor dyes with CellBlox Plus Blocking Buffer to reduce background and to block non-specific binding of NovaFluor labels, PE, and APC tandems observed with macrophages and monocytes.
Spectral signature of NovaFluor Red 800 dye. Normal human peripheral blood cells stained with CD4 Monoclonal Antibody (SK3 (SK-3)), NovaFluor Red 800 were used for analysis. Data were acquired on a 5-laser Cytek Aurora Flow Cytometer. Signature was normalized to the peak intensity and corrected by removal of the negative-population MFI.
NovaFluor 800 dye is brighter than FITC and APC and compatible with Brilliant dyes. Human PBMC were stained with CD8a Brilliant Violet™ 480 and CD4 (clone SK3) APC (left), CD4 (clone SK3) FITC (middle), or CD4 (clone SK3) NovaFluor Red 800 dye. Viable cells in the lymphocyte gate were used for analysis, and CellBlox Plus Blocking Buffer was used when staining with NovaFluor Red 800 dye. Data was collected on a 5-laser Cytek Aurora Flow Cytometer.
NovaFluor dyes are built using Phiton technology and are compatible with both spectral flow cytometry and traditional flow cytometry. NovaFluor dyes exhibit narrow emission spectra and minimal cross-laser excitation, which helps reduce spectral spillover for better marker resolution. In addition, their unique spectral signatures can provide the opportunity to detect additional markers in flow cytometry panels by opening up previously unusable channels.
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
A handy reference poster featuring the broad range of our available dyes and labeling reagents.
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
A tool for selecting the optimal fluorescent dyes for your Invitrogen EVOS cell imaging systems.
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.