Click-iT® Labeling Technologies
Click chemistry is the detection method of choice for samples that would be compromised by direct labeling or antibody-based secondary detection techniques. The click label is small enough to penetrate complex samples easily, and the selectivity and stability of the click reaction provide high sensitivity and low background signal. This gentle sample treatment together with the biocompatible Click-iT® Plus reaction means that detection can be multiplexed with expressed proteins such as GFP, protein labels such as R-PE, and a wide range of organic fluorophores.
Browse Click-iT® assay kits and Click-iT® tools below to find the product that best fits your experiment.
Click-iT® Assay Kits
  • Click-iT® Assay Kits for imaging
  • Click-iT® Assay Kits for HCS
  • Click-iT® Kits for biomolecule capture
  • Click-iT® Kits for flow cytometry
Click-iT® tools
  • Protein and glycan labeling tools
  • Nucleic acid labeling tools
  • Detection tools for standard, low-copper, and copper-free reactions
Find Click-iT® kits and other products by application
Cell proliferation
Measure cell proliferation using imaging or flow cytometry
Apoptosis
Detect late-stage apoptosis using imaging or flow cytometry
Click-iT® technology overview
Click-iT® reactions use azides and alkynes as specific binding moieties in a two-step procedure that first labels and then subsequently detects your molecule of interest. Azides and alkynes can be used interchangeably in “click” reactions, so either one can serve as the tagged substrate or be labeled for the detection step.
Small labels
Azides and alkynes are extremely small, inert, and stable moieties that can be incorporated into biological molecules. They are small enough that nucleotides, sugars, or amino acids can be tagged and still continue to function in the cell and incorporate normally into nucleic acids, complex polysaccharides, or proteins.
Small molecules permit:
  • Cellular loading with minimal disruption
  • Normal incorporation into biomolecules
In a typical Click-iT® labeling step, a cell is supplied with the azide or alkyne moiety in a tagged molecule and it is incorporated into a cellular biopolymer.
Specific detection
Azides and alkynes bind together in a “click” reaction with high specificity to form stable, inert products. We’ve developed both molecules with fluorescent labels attached, such as Alexa Fluor® dyes, to serve as sensitive detection tools that are small enough to penetrate complex samples.
  • Cellular labeling with minimal disruption
  • Specific detection with low background signal
If an azide is incorporated into a cellular protein it can be labeled using an alkyne tagged with an Alexa Fluor® dye. Conversely, if an alkyne is incorporated into a cellular protein it can be labeled using an azide tagged with an Alexa Fluor® dye.
Minimal denaturation
The copper-catalyzed click reaction links an alkyne with an azide to form a stable inert product. The mild reaction conditions of the Click-iT® Plus reagent help preserve the activity of fluorescent proteins, so click reactions can be multiplexed with GFP or R-PE in your detection strategy.
Resources