Click-iT® Assay Kits
![]() |
Click-iT® technology provides a streamlined and simplified workflow for multiple applications in microscope imaging, flow cytometry, and high-content analysis or screening. We’ve developed a series of optimized assay kits designed to generate reliable results, including kits to measure cell proliferation, new protein synthesis, RNA synthesis, lipid peroxidation, and apoptosis using TUNEL assays. Additionally, Click-iT® technology can be used as a sensitive tool for molecular capture, and we offer kits to capture nascent proteins and RNA. The topics below will assist you in selecting the right kit based on your assay platform and application. |
Click-iT® Assay Kits
Assay kits for imagingClick-iT® Assay Kits for imaging provide a robust workflow with a selection of detection options to allow for multiplexing and provide a broader biological context for your results. Imaging kits are optimized for microscopy and offered with a range of Alexa Fluor® dyes for detection. Assay kits for HCSClick-iT® HCS Assay kits are optimized for higher throughput and provide reagent for multiple microplates in each kit. Not all imaging kits are provided in a high throughput format. |
Proliferating HeLa cells detected using the Click-iT® Plus EdU Alexa Fluor® 488 Imaging Kit. |
Assay kits for flow cytometryClick-iT® Assay Kits for Flow Cytometry provide the most convenient to measure cell proliferation by tracking new DNA synthesis. Cell proliferation assays are offered with a range of Alexa Fluor® dyes for detection to allow for multiplexing with common fluorophores, including R-PE and R-PE tandems, as well as fluorescent proteins. Kits for biomolecule captureWhen Click-iT® technology is used for molecular capture, it enables the enrichment of newly synthesized RNA or protein species that can be used in subsequent gene expression analysis or protein sequencing. |
![]() Compatibility of Click-iT® Plus EdU reaction with R-PE tandem conjugate (R-PE Cy®7) fluorescence. |
Ordering information
| Product | Quantity | Cat. No. |
|---|---|---|
| Click-iT® assay kits for cell proliferation | ||
| Click-iT® EdU Pacific Blue™ Flow Cytometry Assay Kit | 1 kit, 50 tests | C10418 |
| Click-iT® EdU Alexa Fluor® 488 Flow Cytometry Assay Kit | 1 kit, 50 tests | C10425 |
| Click-iT® EdU Alexa Fluor® 488 Flow Cytometry Assay Kit | 1 kit, 100 tests | C10420 |
| Click-iT® EdU Alexa Fluor® 647 Flow Cytometry Assay Kit | 1 kit, 50 tests | C10424 |
| Click-iT® EdU Alexa Fluor® 647 Flow Cytometry Assay Kit | 1 kit, 100 tests | C10419 |
| Click-iT® Plus EdU Alexa Fluor® 488 Flow Cytometry Assay Kit | 1 kit, 50 tests | C10632 |
| Click-iT® Plus EdU Alexa Fluor® 488 Flow Cytometry Assay Kit | 1 kit, 100 tests | C10633 |
| Click-iT® Plus EdU Alexa Fluor® 647 Flow Cytometry Assay Kit | 1 kit, 50 tests | C10634 |
| Click-iT® Plus EdU Alexa Fluor® 647 Flow Cytometry Assay Kit | 1 kit, 100 tests | C10635 |
| Product | Quantity | Cat. No. |
|---|---|---|
| Click-iT® kits for nascent protein capture | ||
| Click-iT® Protein Enrichment Kit, for click chemistry capture of azide-modified proteins | 1 kit | C10416 |
| Click-iT® kits for nascent RNA capture | ||
| Click-iT® Nascent RNA Capture Kit, for gene expression analysis | 1 kit | C10365 |
Resources
Learn more
|
BioProbes® Journal articles
Scientific poster
|