From Now until September 30, Find Exclusive Savings on Ambion® Essentials RNA Products*
Receive a $5 Starbucks Card with the purchase of any two Ambion® Essentials products. Restrictions apply.*
  • Simply use promo code xxxxxx when placing your order.
Ambion® Essential Products
With RNA essentials, you can expect our Ambion® RNA expertise, packaged in products that offer the very best for the stabilization, isolation, and analysis of RNA.
Ambion® RNAlater® Solution
Ambion® RNAlater® Tissue Collection: RNA Stabilization Solution is an aqueous, nontoxic tissue storage reagent that rapidly permeates tissues to stabilize and protect cellular RNA.
  • Single reagent immediately inactivates RNases and stabilizes RNA within tissues or cells
  • Eliminates the need to grind and freeze samples in liquid nitrogen or rush samples back to the lab freezer
  • Stabilizes RNA for 1 day at 37°C, 1 week at 25°C, 1 month at 4°C, or indefinitely at –20°C
Ambion® RNaseZAP®
Ambion® RNaseZap® is a surface decontaminant that destroys RNases on contact. 
  • Removes RNase contamination from glass and plastic surfaces
  • Proven effective at removing high concentrations of dried-on RNase A
  • Ideal for cleaning work surfaces, pipettes, and equipment that must be RNase-free
Ambion® DNAZap™
Ambion® DNAZap™ consists of two solutions that are innocuous when used alone, but become a potent nucleic acid degrading solution when mixed. 
  • Completely and instantaneously degrades contaminating DNA and RNA from surfaces on contact—at the level of PCR sensitivity
  • Ideal for cleaning PCR tubes, PCR machine surfaces, pipettes, lab benches, lab equipment, and microfuge tubes
Ambion® Tips and Tube Products
Nuclease-Free Pipette Tips
  • Ambion® barrier tips prevent cross-contamination of samples by means of an aerosol filter. They are ideal for handling RNA, PCR applications, and for working with radioisotopes or toxic chemicals.
Nuclease-Free Microfuge, Conical, PCR Tubes and Caps
  • Each lot of tubes is subjected to rigorous testing and is certified to be nuclease-free. 
Terms & Conditions
 FOR RESEARCH USE ONLY. NOT INTENDED FOR ANY ANIMAL OR HUMAN THERAPEUTIC OR DIAGNOSTIC USE.
.
*This promotion is open to customers in the US and Canada who purchase two qualifying Ambion® Essentials products in a single purchase order. Offer applies to qualifying orders received by Life Technologies no later than September 30, 2011 or until gift supplies are depleted, whichever comes first.  Limit one Starbucks Card (approximate retail value $5 USD) per customer.  This offer cannot be combined with other discounts or promotions.  Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.  Starbucks is not a participating partner or sponsor in this offer.