![]() | We offer a wide range of antibiotics and antimycotics that can help control or eliminate cell culture contamination. |
Find the right cell culture antibiotics for your experiment
Browse the catalog
Use the link below to see all antibiotic and antimycotic products in the catalog, including selection antibiotics. The 'Shop all' view allows you to apply product filters to narrow the displayed list.
Selected cell culture antibiotics
Keep your irreplaceable cultures contamination free
Researchers are typically concerned about two types of cell culture contamination: the contamination of cell cultures with microbiological organisms and the contamination of one cell line with another. Both types of contamination are extremely prevalent, and the possibility of contamination—and the impact on results—cannot be underestimated. The decision to use antibiotics and antimycotics to prevent contamination should be based on the individual researcher’s needs and experience.
We manufacture a wide array of antibiotics and antimycotics to control contamination arising from bacteria, fungi, mycoplasma, and yeast. While researchers may attempt to eliminate or control contaminations with antibiotics or antimycotics, it should be noted that these substances can be toxic to certain cell lines. Before using, it is essential to first perform a dose-response test to determine the level at which toxicity begins.
Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Shop for everyday lab essentials to power your transfection steps
We've made it easier for you to quickly find and buy products for every step in your transfection workflow.
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Media Formulation Tool
Find the right Gibco media formulation for DMEM, DMEM/F-12, MEM, and RPMI-1640 media.
Related products
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products
For Research Use Only. Not for use in diagnostic procedures.