Ampicillin Sodium Salt is a semi-synthetic penicillin derived from the penicillin nucleus, 6-amino-penicillanic acid. It causes cell death by inhibiting cell wall biosynthesis. Resistance to ampicillin is mediated by β-lactamase cleavage of the β-lactam ring (bla gene).
Application(s): Selection of resistant bacteria.