Fosmidomycin
Fosmidomycin, the phosphonate antibiotic FR-31564, is an effective antimalarial agent that functions by blocking a mevalonate-independent pathway of isoprene synthesis.
Search Thermo Fisher Scientific
Fosmidomycin, the phosphonate antibiotic FR-31564, is an effective antimalarial agent that functions by blocking a mevalonate-independent pathway of isoprene synthesis.
Note: You clicked on an external link, which has been disabled in order to keep your shopping session open.