Neomycin
Neomycin Sulfate is an aminoglycoside antibiotic that inhibits translation by binding to the small subunit of prokaryotic ribosomes. Inhibits PLC, phosphatidylcholine-PLD activity, and angiogenin-induced angiogenesis.
USP grade, nonsterile