Informational:Informational:Save up to 31% on your order of cell culture products. View promotion details »
Gibco™
Penicillin-Streptomycin (5,000 U/mL)
Catalog number: 15070063
Gibco™
Penicillin-Streptomycin (5,000 U/mL)
Catalog number: 15070063
Catalog Number
15070063
Unit Size
100 ml
Price (USD)
Price: 25.33
Online price: 23.65
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
15070063100 ml
Price: 25.33
Online price: 23.65
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. Penicillin was originally purified from the fungi Penicillium and acts by interfering directly with the turnover of the bacterial cell wall and indirectly by triggering the release of enzymes that further alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to the 30S subunit of the bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria. This solution contains 5000 units/mL of penicillin and 5000 μg/mL of streptomycin.
We offer a wide range of antibiotics and antimycotics in both powder and liquid formats. See the complete list, or find products for:
Contamination control
Eukaryotic and bacterial selection
See recommended working concentrations for selection antibiotics.
Learn more about the use of antibiotics and antimycotics in cell culture and review guidelines for decontaminating cultures.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration
100 X
Culture Type
Mammalian Cell Culture, Insect Cell Culture
Form
Liquid
Product Type
Antibiotic
Shelf Life
12 Months
Sterility
Sterile-filtered
For Use With (Application)
Prevention of Cell Culture Contamination
Shipping Condition
Dry Ice
Contents & Storage
Storage conditions: -5°C to -20°C
Shipping conditions: Dry ice
Shelf life: 12 months from date of manufacture
Figures
Documents & Downloads
Certificates
    Frequently asked questions (FAQs)
    Citations & References
    Search citations by name, author, journal title or abstract text