Gibco™
Polymyxin B Sulfate
Catalog number: 21850029
Gibco™
Polymyxin B Sulfate
Catalog number: 21850029
Catalog Number
21850029
also known as 21850-029
Unit Size
25 MU
Price (USD)
Price: 510.00
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
21850029
also known as 21850-029
25 MU
Price: 510.00
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Polymyxin B Sulfate is produced by Bacillus polymyxa. Polymyxin B Sulfate works by binding to the cellular membrane and interfering with its permeability. Gibco™ Polymyxin B Sulfate is used for the prevention of bacterial contamination of cell cultures. This antibiotic is highly active against gram-negative bacteria. The recommended working concentration is 100 units/ml. We offer a variety of antibiotics and antimycotics for cell culture applications.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration
100 Units/mL
Form
Powder
Product Type
Antibiotic
Shelf Life
24 Months
Sterility
Non-sterile
For Use With (Application)
Bacterial Selection
Shipping Condition
Room Temperature
Contents & Storage
Storage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Figures
Documents & Downloads
Certificates
    Frequently asked questions (FAQs)
    Citations & References
    Search citations by name, author, journal title or abstract text