Gibco Ham's F-10 Nutrient Mixture Media is formulated for serum-free growth Chinese Hamster Ovary (CHO) cells and the growth of various mammalian cell lines with serum supplementation.
Rely on Gibco Ham's F-10 Nutrient Mixture, formulated for superior quality, greater reliability and improved consistency to give you better overall efficiency and more robust data.
Need a custom F-10 Nutrient Mixture formulation?
Improve cell viability and growth with GlutaMAX media and supplement
While results may vary depending on the cell line, using Gibco GlutaMAX-I Supplement in place of L-glutamine in cell culture can improve cell viability and growth, potentially increasing productivity levels. Increase media stability, minimize toxic ammonia build-up, and maximize cell performance. Get the most from your cell cultures with GlutaMAX media and supplement.
Learn more about:
Mammalian cell culture with Gibco classical media products
For all your additional mammalian cell culture needs, see products in these basal media categories: