showcase.par.10590.image.0.0.1
  • Lower endotoxin and hemoglobin levels
  • Ideal for use with sensitive cells
Order Gibco® Certified FBS now:
Certified FBS
Certified FBS undergoes special biochemical and hormonal profile and bacterial testing.
  • Endotoxin ≤5 EU/ml
  • Hemoglobin ≤10 mg/dl
Heat Inactivated, Certified FBS
Heat Inactivated, Certified FBS is well suited for immunological work.
  • Heated for 30 minutes at 56° C with mixing to inactivate complement
  • Same endotoxin and hemoglobin profile as Gibco® Certified FBS