Gibco Sera Product Categories - Science Research Areas
  Standard Performance Performance Plus Secure Specialty
Cells Robust cell lines Common cell lines Broad range of cells including sensitive cell lines Cell culture requiring lowest viral risk Sensitive cells
Endo Specs / Product Attribute Spec <50 EU/mL  Spec <10 EU/mL  Spec <5 EU/mL plus biochemical and hormonal testing  </=10 EU/mL*
Sourced from BSE-free region
Specialty process of assay qualified
*for fetal bovine and donor bovine sera
Science Research Area Standard Performance Performance Plus Secure Specialty
Cancer Research          
Cell Banking          
Cell Signaling Pathway Research          
Cytogenetics          
Drug Screening          
Gene or Cell Therapy          
Immunology          
IVD Markers          
Molecular Biology          
Neurobiology Research          
Protein Expression          
Stem Cell Research          
Tissue Engineering          
Toxicology Research          
Virus / Plasmid Expression          
Disclaimer: Sera selections are to serve merely as a general recommendation. Please note that each research area listed may include a specific application that requires an alternative sera selection. Each user remains solely responsible to ensure that the selected sera is appropriate and suitable for his or her purpose.