Gibco® horse serum is donor-herd collected, processed, and manufactured in New Zealand. Each lot of horse serum is tested for its ability to support the growth of Sp2/O-Ag14 (murine myeloma) cells in control medium containing the test lot of serum. Results are compared to parallel results obtained using control growth medium containing a previously characterized reference serum.
  • EIA (Equine Infectious Anemia) tested
Horse serum is available in both a heat inactivated and non-heat inactivated format.  HI serum is heated for 30 minutes at 56°C while mixing to inactivate the complement.
Common Uses
  • 10% horse serum could be used to support an in vitro antibody response to SRBC in place of FBS1
  • Used in the production of C2 heterokaryons2
  • Primary Neuronal Culture
References
  1. Miller Levy, E., The ability of horse serum to support…, Journal of Immunological Methods Volume 36, Issue 2, 27 August 1980, Pages 181-183
  2. Production of Heterokaryons. Stanford University Protocols.