Sheep
Gibco Sheep serum is collected from animals younger than 1 year old. It is processed, and manufactured in New Zealand. Gibco Sheep serum is tested specifically for cytotoxicity. Test samples are examined for the ability to support the attachment and proliferation of three adherent cell lines. The test and reference cultures are monitored for evidence of nutritional deficiency, abnormal morphology, or cytotoxicity. Sheep serum is also available in custom formats; call your local representative for more details
Common Uses
  • Used in antibody production1
References
  1. Sun, T., Evaluation of a vaccine against mannheimia haemolytica and pasteurella multocida in sheep, College of Agriculture and Life Sciences Honors These, Issue 7/02/09