Fetal bovine serum (FBS) for standard applications
Value FBS is appropriate for standard research applications, and each lot is subjected to up to 50 quality specification tests including 9CFR virus testing, endotoxin, and performance. Manufactured using triple 0.1 micron filtration.*
Product specifications | Value Plus FBS– United States |
Value FBS– Mexico/Central America |
---|---|---|
Endotoxin | ≤10 EU/mL (typically ≤5 EU/mL) |
≤50 EU/mL (typically ≤10 EU/mL) |
Performance (growth) | ||
9CFR virus testing | ||
Filtration | ||
Total protein | 3–5 g/dL | 3–5 g/dL |
Hemoglobin | ≤25 mg/dL | ≤25 mg/dL |
Mycoplasma | ||
pH (6.9–7.8) | ||
Osmolality (280–340 mOsm/kgH20) | ||
Origin | United States | Mexico/Central America |
Base Cat. Nos. | 26140, 16140, A31605, A38401 | 10437, 10438, A31606, A38402 |
Buy 50 mL, 100 mL, 500 mL, or 1 L | Order now | Order now |
Buy 50 mL, 100 mL, 500 mL, or 1 L, heat-inactivated | Order now | Order now |
= testing is performed. Gamma-irradiated Value FBS is available upon request. |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.