To prolong the viability of cells in culture and stimulate growth, supplements of amino acids can be added to media.  In addition, adding the supplement adds nutrients and reduces biosynthetic burden on cell in vitro.  Prepared in distilled water. The non-essential amino acids in this solution are 100X the concentration in a MEM-alpha Medium.
For Research Use Only. Not for use in diagnostic procedures.