CHO (Chinese hamster ovary) Medium is a chemically-defined, protein free medium optimized for the growth of CHO cells (including CHO-S cells) and the production of recombinant proteins in suspension culture. It contains no components of animal origin and is formulated without L-glutamine, providing added stability.