Yeast nitrogen base with amino acids is a satisfactory nitrogen source for general growth media. Contains all essential nutrients required for the growth of yeasts except carbohydrate. Addition of a carbon source is required.