Hank's Balanced Salt Solution
The essential function of a balanced salt solution is to maintain pH and osmotic balance as well as provide your cells with water and essential inorganic ions. Gibco HBSS is manufactured in our state-of-the-art cGMP, ISO certified facilities, so you can rely on the highest quality and consistency.
Gibco HBSS is available in multiple formulations and formats to meet your specific research needs, including calcium- and magnesium-free versions for washing cells prior to trypsinization.