Reliable dissociation for adherent cell culture
Gibco trypsin and alternative cell dissociation products are ideal for use with tissues and cell monolayers. Gibco trypsin and TrypLE reagent come in a wide variety of formats to meet the diverse needs of researchers performing adherent cell culture.
Trypsin and alternative cell dissociation reagents
Fast, general purpose | Gentle & convenient for research use | Gentle & convenient for bioproduction/ industrial applications | Gentle & fast for strongly adherent cells | Specialized for neural & pluripotent stem cell dissociation | |
---|---|---|---|---|---|
Trypsin | TrypLE Express (1X) | TrypLE Select (1X) | TrypLE Select (10X) | StemPro Accutase | |
Learn more › | Learn more › | Learn more › | Learn more › | Learn more › | |
Source origin | Animal origin (porcine) | Animal origin-free | Animal origin-free (dedicated AOF machinery) | Animal origin-free (dedicated AOF machinery) | Serum free, animal origin (invertebrate) |
Storage temperature | Frozen (–5 to –20° C) | Ready to use (room temp. stable) | Ready to use (room temp. stable) | Ready to use (room temp. stable) | Refrigerated (2–8°C) |
Higher cell viability | ✓ | ✓ | ✓ | ✓ | |
Inactivation method | Requires Trypsin inhibitors / neutralizers | Inhibition by dilution (no inhibitors needed) | Inhibition by dilution (no inhibitors needed) | Inhibition by dilution (no inhibitors needed) | Inhibition by dilution (no inhibitors needed) |
Available packaging | 100 mL 50 mL 100 g powder | 100 mL 500 mL 5 L bags | 100 mL 500 mL | 100 mL 500 mL | 100 mL |
Select products | Order 1x, with phenol red Order 1x, no phenol red | Order 1x, no phenol red | Order 10x, no phenol red | Order |
Other dissociation products | Tissue dissociation |
GMP manufacturing
Gibco cell culture products are manufactured in facilities that are compliant with current good manufacturing practices (GMP) for medical devices and adheres to a robust quality management system.
Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Media Formulation Tool
Find the right Gibco media formulation for DMEM, DMEM/F-12, MEM, and RPMI-1640 media.
Related products
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products
For Research Use Only. Not for use in diagnostic procedures.