Meets USP Monograph for Water for Injection packaged in bulk for commercial use elsewhere, 0.1 micron filtered.
The Certificate of Analysis Provides the Following Parameters:
Parameter Limits Units
Endotoxin Testing <= 0.25 EU/mL
Conductivity Measurement <= 5.0 us/cm
Osmolality >= 0 to <= 20 mOsm/Kg
Oxidizable Substances Meets USP requirements  
pH >= 4.0 to <= 7.5  
Sterility Testing Negative