Protein Expression Medium (PEM)
Protein Expression Medium (PEM) is an animal-origin-free medium recommended for the growth of transfected PER.C6® cells for the production of recombinant proteins. Supplementation with L-glutamine or GlutaMAX™-I Supplement is required. Each lot of PEM is tested for performance in cell culture.
- Efficient production of recombinant proteins in PER.C6 cells
- Supports the large-scale high-density culture of transfected PER.C6 cells in shake flasks, roller bottles and bioreactors
- Formulated without L-glutamine for maximum stability
- Supports parental PER.C6, HEK 293, and HeLa cell growth in suspension culture, as well as transfected PER.C6 cells for protein expression applications