Search
Primary human melanocyte cells are shown to proliferate in our Primary Human Melanocytes (HEMa) Cell Culture Systems. Our Melanocyte Cells cultures are optimized to synthesize melanocyte cells, which are derived from the melanogenesis process.
HEMa cells grown in Medium 254 (M-254-500) with the addition of Human Melanocyte Growth Supplement (HMGS). First culture after thaw.
![]() Day 5 - HEMa | ![]() Day 9 - HEMa | ![]() Day 14 - HEMa |
Primary cell culture support center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
For Research Use Only. Not for use in diagnostic procedures.