Synthetic genes for creating challenging constructs and protein expression studies
Fast turnaround times and sequence accuracy
Introduction to Gateway® recombination cloning technology
View video
Speed through everyday cloning
Find Invitrogen cloning products
Highly-efficient, vector independent systems for simultaneous and seamless assembly:
GENEART® Seamless Cloning and Assembly Technology
Up to four DNA fragments plus a vector totaling up to 13 kb in length
GENEART® High-Order Genetic Assembly System
Up to ten DNA fragments plus a vector totaling up to 110 kb in length
GENEART® Site-Directed Mutagenesis System
Quick, highly-efficient in-vitro site-directed mutagenesis of plasmids up to 14 kb