Ultimate ™ ORF Clones BAC and PAC Clones Yeast Deletion Clones Yeast-GFP Clones
Number of clones in collection ~17,000 ~3 million ~20,000 ~4100
Available species Human and Mouse Human, Mouse and Rat Yeast Yeast
Sequence information Full-insert sequenced not verified   PCR verified to contain an insert of the correct length
Expression tested No No No No
Gateway™ Technology compatible Yes, in pENTR™ 221 No No No
Amino acid guarantee Yes, 100% match No No No
QC parameters Growth, phage tested and end sequenced prior to shipping Growth and phage tested Growth and phage tested Growth and phage tested
Applications Function/ Expression studies Screening genomic DNA Function/ Expression studies Localization studies
Cat. No.  (all catalog numbers need a specific clone ID to process the order) HORF01 (human), MORF01 (mouse) Various 95400 95700