Competent Cells by Strain
Obtaining the right clone shouldn't be the time-intensive step in your research. That's why each E. coli strain is specially engineered to keep you moving ahead. High transformation efficiencies - ranging from 1 x 106 to 3 x 1010 - ensure that you produce representative cDNA and genomic libraries every time.