strand issue 1
In This Issue
gene synthesis  
 Featured Video
Synthetic Biology in a Nutshell
Enjoy this TEDx presentation with Dr. Ralf Wagner of Life Technologies....read more>>
New Tools: GeneArt® Precision TALs: Precise Genome Engineering
GeneArt® Precision TALs provide custom DNA-binding proteins for accurate DNA targeting ...read more>>
NewCloning Tools  
Tips & Tricks
Top 5 Gateway® Cloning Tips & Tricks ..read more>>
bioinformatics  
The Future of Bioinformatics
Test drive the future of bioinformatics
Vector NTI® Express software supports the future of bioinformatics and synthetic biology with a plug-in architecture to consistently deliver new tools...read more>>
resources  
Synthetic Biology Resources
Keep up to date with the latest synthetic biology products and applications...read more>>
Did you know?
Life Technologies sequencer identifies European E. coli
What can SynBio do for you? In 2011, the Life Technologies ION PGM Sequencer was used to resolve a major E. coli health crisis. read more>>
Upcoming Events
Come see us at the upcoming Trade shows & Conferences...see where>>
Q&A
Bioinformatics
Learn the latest in our bioinformatics portfolio with Raymond Tecotzky. read more>>
Hot from Social
Recombination-Based DNA Assembly and Mutagenesis Methods.....read more>>
Featured Video
Synthetic Biology in a Nutshell
Enjoy this TEDx presentation with Dr. Ralf Wagner of Life Technologies.
Dr. Wagner was one of the first to realize the enormous scientific and commercial potential of synthetic biology. He successfully converted that scientific and technical vision into a sound business model supplying gene optimization, DNA synthesis and subcloning to the 20 largest pharmaceutical companies in the world. In addition to his work at GeneArt®, Ralf also manages the Molecular Microbiology and Gene Therapy unit at the University of Regensburg. Some people decide to build a better mouse trap. Others, a better mouse.
GeneArt® Precision TALs: Precise Genome Engineering
GeneArt® Precision TALs provide custom DNA-binding proteins for accurate DNA targeting and precise genome editing. Other technologies limit the choice of targets, but with GeneArt® Precision TALs, you can target any locus in the genome, and this increase in engineering accuracy allows you to develop better solutions for healthcare, agriculture, energy, and bioproduction research.
Start editing
Tips & Tricks
Top 5 Gateway® Cloning Tips & Tricks
gateway cloning  
  1. Recovering an entry clone from an expression construct. The efficiency of the BP reaction can be improved by linearizing the expression construct. The BP Clonase® works more efficiently when, at least, one of the substrates is a linear fragment.
  2. Improve the chances to obtain PCR amplified fragments for BP reactions. Relatively short template specific sequence (12-14 mer) improves the chances of amplification success without compromising specificity.
  3. Use destination vectors and entry clones with the same selection marker. It can be performed by linearizing the entry clone with one or more restriction enzymes that do not cut within the insert sequence.
  4. Give a second chance to failed LR or BP reactions. Do not stop the reaction with proteinase K. Instead, after transformation, keep the tube at room temperature and re-transform the following day if necessary.
  5. Speed up the process. Use One Shot® Mach1™-T1R Chemically Competent E. coli cells. Colonies are clearly visible after eight hours of plating. Transformants to be selected on ampicillin plates do not need to be recovered before plating.
Have tips and tricks you use in your lab?  Send us your tips
Test drive the future of bioinformatics
Vector NTI® Express software supports the future of bioinformatics and synthetic biology with a plug-in architecture to consistently deliver new tools and functionality. Life Technologies can provide a customized Vector NTI® Express software quote to meet your needs.
Receive a custom quote
Learn the latest in our bioinformatics portfolio with Raymond Tecotzky, product manager of Vector NTI® software.
Question: What’s the difference between the Vector NTI Advance® and Vector NTI® Express software suites?
Answer: Both the Vector NTI Advance® and Vector NTI® Express software suites allow you to manage, view, analyze, transform, share, and publicize diverse types of molecular biology. The Vector NTI Advance® sequence analysis suite has been around for more than 20 years, while the Vector NTI® Express software is a newer, even faster, simpler, and easier-to-use version of Vector NTI® software. Vector NTI® Express software is also designed to run on both PC and Mac® platforms. You can find a full list of Vector NTI® software features here.
Have questions to ask us?  Send us your questions
*Your question may be included in a future edition of The Strand.
Upcoming Events
FOR RESEARCH USE ONLY. NOT INTENDED FOR ANY ANIMAL OR HUMAN THERAPEUTIC OR DIAGNOSTIC USE.
© 2012 Life Technologies Corporation. All rights reserved. The trademarks mentioned herein are the property of Life Technologies Corporation or their respective owners. Mac® is a registered trademark of Apple, Inc. In compliance with federal regulations, we hereby disclose that this email communication is for commercial purposes.