Strand Newsletter
In This Issue
gene synthesis  
Featured Video
Learn more about precise genome editing....watch video>>
New Tools: Enhanced online ordering of GeneArt® Gene Synthesis and Subcloning Services
Thanks to your feedback, Life Technologies has greatly enhanced online ordering for our gene synthesis and subcloning services....read more>>
Algae Engineering  
Algae Engineering
Looking for quality algae engineering? ..read more>>
resources  
Synthetic Biology Resources
Keep up to date with the latest synthetic biology products and applications...read more>>
q&a  
Precise Plant Genome Editing
Learn about Precision TALs for plants from Reed Hickey, product manager... read more>>
Did you know?
Synthetic Clonal Reproduction Through Seeds
Synthetic Clonal Reproduction
The first study on cloning plants at the seed stage was published in February 2011.Read more>>
Upcoming Events
Come see us at the upcoming Trade shows & Conferences...see where>>
Hot from Social
Rewriting the Script.....read more>>
Featured Video
Learn more about precise genome editing
Precision TALs were derived from Xanthomonas, a genus of plant pathogens. These pathogens specifically recognize the DNA of the host with a protein containing 17–18 repeats of approximately 34 amino acids each. Enjoy this video about Precision TALs, which have been shown to be effective in a broad range of organisms, including plants, algae, and fungi.
GeneArt® Gene Synthesis and Subcloning Services
Enhanced online ordering of GeneArt® Gene Synthesis and Subcloning Services
Thanks to your feedback, Life Technologies has greatly enhanced online ordering for our gene synthesis and subcloning services. Fewer working steps, combined with intelligent features, results in quicker and easier handling.
Among other improvements, you will benefit from:
  • One step to enter all information for a sequence that does not need any edits
  • Automated in silico cloning that immediately delivers a vector map and gb file of your cloned gene construct
See how easy it is
Algae Engineering
Algae Engineering    
Looking for quality algae engineering?
Released in late 2011, GeneArt® Algae Engineering Kits for Chlamydomonas reinhardtii and Synechococcus elongatus are the first commercially available genetic modification and expression systems for photosynthetic microalgae.
Find out more »
Development of synthetic biology kits for microalgae
In this scientific poster from 2011, our R&D team gives you a look at the challenges and discoveries behind GeneArt® algal product systems.
Read more »
Precise Plant Genome Editing
Q&A  
Learn about Precision TALs for plants from Reed Hickey, product manager.
Q: Why are GeneArt® Precision TALs such a good fit for plant genome engineering?
A: TAL is the first technology of its kind that not only allows researchers to target any locus in the genome, but also offers a clear path for commercial use of their discoveries through the Two Blades Foundation. TAL is also a plant-based technology, and has been effective in organisms like algae, fungi, and other plants. Native TAL Fok1 is the best ordering option for plant science research.
Contact us for more on plant editing
Have questions to ask us?  Send us your questions
*Your question may be included in a future edition of The Strand.
Upcoming Events
plant engineering    
Upcoming webinar on plant engineering
Learn more about GeneArt® Precision TALs, as well as Gateway® and GeneArt® cloning technologies for plant science research. Please email us if you're interested in attending one of our free live webinar sessions on October 9 and 11, or if you'd like to view a recorded version click here.
Hot from Social
social  
Rewriting the Script
"...many genetic diseases arise because people carry broken or dysfunctional versions of one gene or another. If there were a way to add a properly working copy of those genes back in, the associated diseases should go away, or at least get a whole lot better."
FOR RESEARCH USE ONLY. NOT INTENDED FOR ANY ANIMAL OR HUMAN THERAPEUTIC OR DIAGNOSTIC USE.
© 2012 Life Technologies Corporation. All rights reserved. The trademarks mentioned herein are the property of Life Technologies Corporation or their respective owners. Mac® is a registered trademark of Apple, Inc. In compliance with federal regulations, we hereby disclose that this email communication is for commercial purposes.