BindIt Software
Create and store protocols in a PC database using Thermo Scientific BindIt Software for Thermo Scientific KingFisher instrument systems. Once a protocol has been created, the protocol can either be transferred to your KingFisher instrument memory or executed directly from the software. Based on a step list, the parameters for the active step are shown on the screen. All steps have default parameters which can be changed according to the demands of the application. Alternatively, download and execute protocols from our extensive library.
Features of BindIt Software
  • Compatible with open-platform KingFisher instrument systems
  • Allows specific plates and reagents to be defined in the plate layout
  • Generates status reports that include run log, plate layout, and step parameters
  • Enables the KingFisher Flex to interface with liquid handling, robotics, and plate stacking instruments, providing a fully automated solution and the highest possible throughput
Already have a KingFisher Instrument and BindIt Software?
Download BindIt Software Upgrade
Protocols for BindIt Software
All KingFisher instruments come with dedicated BindIt PC software, which includes optimized protocols for a variety of Invitrogen and Thermo Scientific purification kits and reagents. Here, we provide users with access to download new protocols for free as they become available. Choose protocols designed for different genomic nucleic acid, DNA, RNA, and protein purification procedures and reagent kits from various manufacturers.