KingFisher Instrument Plastics
Optimize performance using Thermo Scientific KingFisher Systems Plastics, specifically designed for use with the KingFisher System and ideal for magnetic particle processing because of their low binding affinity for biomolecules. The specially designed, disposable tip combs and microplates, which are made of polypropylene, enable excellent recovery of magnetic beads.
Plates and tip combs used with KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto, including new and temporary plastics to support global demand for SARS-CoV-2 testing.
 Brochure: SARS-CoV-2 support and solutions KingFisher instruments and MagMAX isolation kits
 KingFisher plastics for SARS-CoV-2 testing
 Application note: MagMAX Viral/Pathogen kit protocol changes to enable increased SARS-CoV-2 testing throughput