Bacteria is present everywhere and as research progresses, we are furthering our understanding of the important role that bacteria play in human health. The study of the microbiome is influencing our understanding of areas such as diabetes, malnutrition, obesity, and women’s health.
Not all bacterial cells are the same, and gram positive samples can be especially challenging to lyse. We have developed a variety of product formats to address all the bacterial cell types from those easy-to-lyse to the more difficult gram positive cells.
Which Genomic DNA Extraction From Bacteria Kit is right for you?
  Order now Order now Order now Order now
  Most cost effective Fast isolation of gDNA from bacterial cells High-yield, high-purity gDNA in a plate format Rapid and automated extraction of DNA
  DNAzol™ Reagent PureLink™ Microbiome DNA Purification Kit PureLink™ Pro 96 Genomic DNA Purification Kit MagMAX™-96 DNA Multi-Sample Kit
Cell input Up to 3x107 cells Up to 2x109 cells Up to 2x109 cells Up to 2x109 cells
Yield Up to 70 µg Up to 25 µg Up to 10 µg 15-30 µg
Isolation method Organic extraction Silica membrane Filter plate Magnetic beads
High-throughput compatible No No Yes Yes
Compatible applications Cloning, qPCR, sequencing Cloning, qPCR, sequencing, genotyping Cloning, qPCR, sequencing, genotyping Cloning, qPCR, sequencing, genotyping
Prep time 10-30 min 45 min 35 min 45 min
Prep size 100 mL 50 preps 4 x 96 preps 50, 96, 500, 2500  preps
Resources
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.
DNA Purification Brochure
RNA Isolation & Purification Brochure
For Research Use Only. Not for use in diagnostic procedures.