DNA extraction from cells is one of the most widely used molecular biology techniques. This has traditionally been a highly repetitive and labor-intensive procedure, requiring significant time investment, and often involving the use of toxic organic chemicals. We have developed purification products that are optimized to provide DNA of maximum yield, purity, and integrity from cultured cells, including cell suspensions using several technology formats; organic extraction, silica membranes, and magnetic beads. These options provide users with flexibility, throughput, and choice along both the product and price continuum.
Which DNA Extraction From Cells Kit is right for you?
  Order now Order now Order now
Order now
  Process the most cells Fast isolation of gDNA from cells High -yield, high-purity gDNA in a plate format Rapid and automated extraction of DNA 
  DNAzol™ Reagent PureLink™ Genomic DNA Mini Kit PureLink™ Pro 96 Genomic DNA Purification Kit MagMAX™-96 DNA Multi-Sample Kit
Cell input Up to 3x107 cells Up to 5x106 cells Up to 5x106 cells Up to 5x106 cells
Yield Up to 70 µg Up to 10 µg Up to 10 µg 15-30 µg
Isolation method Organic extraction Silica membrane Filter plate Magnetic beads
High-throughput compatible  No No Yes Yes
Compatible applications Cloning, qPCR, sequencing Cloning, qPCR, sequencing, genotyping Cloning, qPCR, sequencing, genotyping Cloning, qPCR, sequencing, genotyping
Prep time 10-30 min 15 min 35 min 45 min
Prep size 100 mL 50, 250  preps 4 x 96  preps 50, 96, 500, 2500  preps
PureLink® Genomic DNA Mini Kit  
Higher, more concentrated yields achieved with the PureLink® Genomic DNA Mini Kit.
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.
DNA Purification Brochure
RNA Isolation & Purification Brochure
For Research Use Only. Not for use in diagnostic procedures.