Biological fluid samples, such as plasma, serum, and urine, contain cell-free DNA (cfDNA). cfDNA is used in pathogen detection, and cfDNA targets are being used as biomarkers in oncology research. Isolation of cfDNA is challenging because of its low abundance in typical samples. In pathogen detection, virus particles are also typically in low abundance and therefore are challenging to capture. Thus, large volumes of biological fluids (which can be difficult to handle) are required to obtain sufficient amounts of cfDNA for analysis. We have developed scalable products capable of handling these large volumes while efficiently recovering cfDNA from a broad range of sample types in several format options.
Which DNA extraction kit for plasma, serum, and urine samples is right for you?
  Order now Order now Order now
  Fast isolation of viral nucleic acid Easy-to-use, high-sensitivity Automated format, capable of processing large volumes of plasma
  PureLink™ Viral RNA/DNA Mini Kit PureLink™ Pro 96 Viral RNA/DNA Purification Kit MagMAX™ Cell-Free DNA Isolation Kit
Sample input 500 µL 200 µL 0.5–10 mL
Compatible samples Plasma, serum, cerebrospinal fluid Plasma, serum, cerebrospinal fluid Plasma, serum, urine
Isolation method Silica membrane Filter plate Magnetic beads
High-throughput compatible No Yes Yes
Compatible applications Cloning, qPCR, sequencing, genotyping Cloning, qPCR, sequencing, genotyping Cloning, qPCR, sequencing, genotyping
Prep time 15 min 35 min 24 samples in <1 hr
Prep size 50 preps 4 x 96  preps 50 preps
For Research Use Only. Not for use in diagnostic procedures.