The GeneCatcher™ technology is a novel magnetic bead-based genomic DNA (gDNA) purification technology that is designed to work on a wide range of blood samples including archived or poorly stored blood samples.  The GeneCatcher™ gDNA blood kits allow rapid and efficient extraction of genomic DNA from human blood—without the use of centrifugation (Figures 1 and 2).  The purified DNA is suitable for use in the most sensitive downstream applications, including PCR and restriction enzyme digestion.
GeneCatcher™ Technology  
Figure 1. How GeneCatcher™ technology works. The novel three-step clean-up process of GeneCatcher™ technology enables scalable, single-tube purification of genomic DNA from large (up to 10 mL) blood samples.
GeneCatcher™ Technology  
Figure 2. Rapid processing of large blood volumes using GeneCatcher™ technology.
GeneCatcher™ Purification System
The GeneCatcher™ Purification System offers a method to purify gDNA from large volumes of blood that is robust and reproducible, ensuring complete sample recovery and integrity. The GeneCatcher™ gDNA Blood Kits allow rapid and efficient extraction of genomic DNA (gDNA) from human blood, including archived or poorly stored blood samples (Figure 3). Genomic DNA is extracted from blood samples using the cost-effective, user-friendly magnetic bead–based GeneCatcher™ technology without the use of centrifugation.
Real-time PCR results from GeneCatcher™ purified gDNA  
Figure 3. Purification of gDNA from archived whole blood samples using the GeneCatcher™ gDNA Blood Kit. An 0.8% agarose gel used to evaluate gDNA extractions from archived whole blood samples (blood samples frozen >8 years). Blood samples were split into 2 equal aliquots, and gDNA was extracted using the GeneCatcher™ gDNA Blood Kit and a kit from competitor G. The GeneCatcher™ kit extractions employ magnetic beads, while competitor G’s kit uses a form of the traditional lysis and DNA precipitation method that has been shown to co-purify agents that are inhibitory to downstream applications. I = GeneCatcher™ technology extractions; G = competitor G extractions; L = 1Kb DNA Extension Ladder (Life Technologies), largest band 40 Kb.
Scalable DNA Isolation Ready for Automation
GeneCatcher™ technology has been designed and optimized for scalable and automated genomic DNA isolation from blood samples using the Tecan® Freedom EVO® liquid handling platform (Figure 4A); It requires no centrifugation or filtration steps and uses reagents that will not clog lines or cause vapor pressure buildup. Furthermore, a magnetic rack (separator) allows processing of several samples simultaneously during the magnetic bead-separation steps. To obtain the best results with the GeneCatcher™ gDNA 0.3–1 mL Blood Kit, we recommend using the 24-well Magnetic Separator (Figure 4B). For the GeneCatcher™ gDNA 3–10 mL Blood Kit, we recommend the 50 mL tube Magnetic Separator (Figure 4B). These magnetic separators are compatible with the volumes used in the kit protocols and provide effective magnetic strength from neodymium magnets, which are aligned with plate wells or tubes, respectively.
GeneCatcher™ technology    Figure 4. Ready for automation. (A) The protocol for the GeneCatcher™ gDNA 3–10 mL Blood Kit can be automated on the Tecan® Freedom EVO® platform, whereas the protocol for the GeneCatcher™ gDNA 0.3–1 mL Blood Kit can be automated on the Hamilton STAR and Tecan® Freedom EVO® platforms. (B) The GeneCatcher™ gDNA 0.3–1 mL Blood Kit works well with the 24-well Magnetic Separator and a 24 Deep Well RB Block. The 24-well Magnetic Separator contains 6 neodymium magnets that are perfectly aligned with the wells of a 24-well plate for simple sample processing using magnetic beads.
For Research Use Only. Not for use in diagnostic procedures.