DNAzol® Reagents are complete, ready-to-use reagents for the isolation of genomic DNA. Genomic DNA isolated with DNAzol® Reagents can be used for restriction endonuclease digestion, Southern blot analysis, molecular cloning, and PCR.
DNAzol® Reagent 
  • Designed for use with tissues, cells, or blood
  • Includes a novel guanidine-detergent lysing solution that selectively precipitates DNA from a cell lysate
  • Offers procedures that are completed in 30 to 60 min
  • Provides detailed protocols on the product profile
  • Can isolate genomic DNA from 50 mg of tissue or 1 x 107 to 3 x 107 cells with one milliliter of DNAzol® Reagent
Learn more about DNAzol® Reagent
DNAzol® BD Reagent 
  • Specifically formulated for the isolation of genomic DNA from blood
  • Can isolate genomic DNA from 0.5 mL whole blood with one milliliter of reagent
Learn more about DNAzol® BD Reagent
Plant DNAzol® Reagent 
  • Specifically formulated for the isolation of genomic DNA from plants
  • Can isolate genomic DNA from 0.1 g of plant tissue with 0.3 mL of reagent
Learn more about Plant DNAzol® Reagent
Ordering Information
Sample Type (General) Final Product Product Name Quantity Cat. No.
Cells Genomic DNA DNAzol® Reagent 100 mL 10503-027
Blood Genomic DNA DNAzol® BD Reagent 100 mL 10974-020
Plant samples Genomic DNA Plant DNAzol® Reagent 100 mL 10978-021
For Research Use Only. Not for use in diagnostic procedures.