Viral DNA extraction poses unique challenges for good recovery and detection sensitivity. Capture and lysis of virus particles from very dilute solutions or cell-free samples is the first hurdle. We have developed DNA purification products that are optimized to provide maximum viral DNA yield, purity, and integrity from a broad range of sample types in several format options.
Viral DNA purification from a variety of sample types
High-throughput recovery low abundance of viral DNA Viral DNA purification from a variety of sample types Consistent recovery of low abundance of viral DNA
Order now Order now Order now
  PureLink® Pro 96 Viral RNA/DNA Purification Kit MagMAX™ Pathogen RNA/DNA Kit PureLink® Viral RNA/DNA Mini Kit
Starting Material 200uL cell free sample 50 - 300 uL 500 µL cell free sample
Sample Type Plasma, serum, cerebrospinal fluid Plasma, serum, whole blood, swabs, feces, ear notches, semen, oral fluid, InPouch™ cultures Plasma, serum, cerebrospinal fluid
Protocol Time 30 min 45 min 45 min
Isolation Technology Silica Plate Scalable, flexible format with magnetic beads Fast, convenient silica column
High Throughput Compatible
Number of Preps 384 480 50
Resources
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.
DNA Purification Brochure
RNA Isolation & Purification Brochure
For Research Use Only. Not for use in diagnostic procedures.