Starting with high-quality plasmid DNA is critical to your workflow.
The table below will help you choose the right plasmid DNA maxiprep kit.
Which plasmid DNA maxiprep kit is right for you?
  Order now Order now Order now Order now
  Quick and cost effective Most widely accepted protocol for transfection Rapid purification with integrated lysate clarification filter Rapid purification with integrated lysate clarification filter and precipitator
  GeneJET Plasmid Maxiprep Kit PureLink™ HiPure Plasmid Maxiprep Kit PureLink™ HiPure Plasmid Filter Maxiprep Kit PureLink™ HiPure FP (Filter and Precipitator) Maxiprep Kit
Purity grade Molecular Transfection Transfection Transfection
Total protocol time 60 min 120 min 90 min 60 min
Yield Up to 750 µg Up to 1 mg Up to 1 mg Up to 1 mg
Technology Silica membrane Anion-exchange gravity-flow column Anion-exchange gravity-flow column Anion-exchange gravity-flow column, precipitator
Clarification filter No No Yes, save 30 minutes Yes, save 30 minutes
Prep size 25, 100 preps 25, 50 preps 25, 50 preps 25, 50 preps
Which plasmid DNA maxiprep kit is right for you?
Eliminates the use of chaotropic salts Fast protocol with integrated lysate clarification Fastest protocol for endotoxin-reducted plasmid Fastest protocol. 60 minutes from pellet to purified plasmid Fully Automated, pour culture and click go, <5 min set up
Order now Order now Order now Order now Order now
  PureLink® HiPure Plasmid Maxiprep Kit PureLink® HiPure Filter Plasmid Maxiprep Kit ChargeSwitch® Pro Filter Plasmid Maxi Kit PureLink® HiPure FP (Filter and Precipitator) BenchPro® 2100 Plasmid Purification System
Total protocol time 120 minutes 90 minutes 30 minutes 60 minutes 90 minutes <5 min hand-on time
Starting Point Cell Pellet Cell Pellet 100 mL Bacterial cultures Cell Pellet 125 mL Cell Culture
Yield plasmid DNA 500-800 µg Up to 850 µg Up to 800 µg Up to 850 µg Up to 750 µg
High-throughput compatible No Yes No No No
Technology/
Format
Silica-based spin/vacuum-column Proprietary ChargeSwitch® magnetic beads Proprietary ChargeSwitch® spin/vacuum-column Anion-exchange gravity-flow column Silica-based Spin-vacuum plates
Product size 10, 25 preps 10, 25 preps 10, 25 preps 10, 25 preps 4 preps
Video: How to purify transfection-grade plasmid DNA
Learn how to purify transfection grade plasmid DNA with the Invitrogen™ PureLink™ HiPure Plasmid Filter Maxiprep Kit.
Resources
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.
DNA Purification Brochure
RNA Isolation & Purification Brochure